The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009018.1 | NP_033044.1 | - | 710 | 95 |
GenBank mRNA | BC132028.1 | - | - | 672 | 95 |
BC132032.1 | - | - | 672 | 95 | |
D64162.1 | - | - | 710 | 95 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198193.2 | NP_937836.1 | 5 - 6 | 746 | 95 |
GenBank mRNA | AK012289.1 | - | 5 - 6 | 746 | 95 |
AK131854.1 | - | 7 - 8 | 1049 | 95 | |
AY054973.1 | - | 3 - 4 | 612 | 95 | |
AY056835.1 | - | 3 - 4 | 612 | 95 | |
BC132308.1 | - | 3 - 4 | 649 | 95 | |
FJ594067.1 | - | 6 - 7 | 850 | 95 |