The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010090.2 | NP_034220.2 | 1 - 2 | 477 | 65 |
GenBank mRNA | AK134067.1 | - | 1 - 2 | 478 | 65 |
BC048696.1 | - | 1 - 2 | 463 | 65 | |
L11330.1 | - | 1 - 2 | 432 | 65 |