Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
Mid1
Entrez Gene ID
17318
Gene Name
midline 1
Gene Aliases
61B3-R, DXHXS1141, Fxy, Trim18
Chromosome Location
Chr.X: 169685199 - 169990798 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010797.3 NP_034927.2 - 1517 89
GenBank mRNA Y14848.1 - - 1471 89

View Full Product Details