The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_134065.4 | NP_598826.3 | 2 - 3 | 749 | 87 |
GenBank mRNA | AF353717.1 | - | 2 - 3 | 562 | 87 |
AK034083.1 | - | 2 - 3 | 718 | 87 | |
AK038733.1 | - | 2 - 3 | 724 | 87 | |
AK046185.1 | - | 2 - 3 | 724 | 87 | |
AK160413.1 | - | 2 - 3 | 684 | 87 | |
AK166252.1 | - | 2 - 3 | 690 | 87 | |
BC014708.1 | - | 2 - 3 | 704 | 87 |