The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173372.2 | NP_775548.2 | 9 - 10 | 2273 | 67 |
GenBank mRNA | BC034118.1 | - | 5 - 6 | 1274 | 67 |
BC041684.1 | - | 4 - 5 | 1282 | 67 |