The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175638.3 | NP_783569.1 | 4 - 5 | 1264 | 83 |
GenBank mRNA | AY184228.1 | - | 4 - 5 | 1233 | 83 |
AY187027.1 | - | 4 - 5 | 1189 | 83 | |
AY260059.1 | - | 4 - 5 | 1227 | 83 | |
BC037114.1 | - | 4 - 5 | 1224 | 83 | |
BC043677.1 | - | 4 - 5 | 637 | 83 | |
BC096453.1 | - | 4 - 5 | 1220 | 83 |