The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026807.2 | NP_081083.2 | 1 - 1 | 717 | 122 |
GenBank mRNA | AK004055.1 | - | 1 - 1 | 718 | 122 |