Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Krtap3-3
Entrez Gene ID
66380
Gene Name
keratin associated protein 3-3
Gene Aliases
2310020D06Rik, KAP3.3, KRTAP3.3
Chromosome Location
Chr.11: 99550132 - 99550863 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025524.2 NP_079800.1 1 - 1 642 133
GenBank mRNA AK009052.1 - 1 - 1 133
AK009410.1 - 1 - 1 644 133
AK010142.1 - 1 - 1 643 132

View Full Product Details