The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011653.2 | NP_035783.1 | 4 - 4 | 1464 | 123 |
GenBank mRNA | AK077529.1 | - | 4 - 4 | 1475 | 122 |
AK145404.1 | - | 4 - 4 | 1513 | 123 | |
AK146983.1 | - | 4 - 4 | 1513 | 123 | |
AK161783.1 | - | 2 - 2 | 1060 | 123 | |
AK164335.1 | - | 4 - 4 | 1515 | 123 | |
AK167687.1 | - | 4 - 4 | 1513 | 123 | |
AK168762.1 | - | 4 - 4 | 1513 | 123 | |
AK169610.1 | - | 4 - 4 | 1514 | 123 | |
BC056169.1 | - | 4 - 4 | 1475 | 123 | |
BC083343.1 | - | 4 - 4 | 1488 | 123 | |
BC083344.1 | - | 4 - 4 | 1483 | 123 | |
BC083345.1 | - | 4 - 4 | 1498 | 123 | |
BC085256.1 | - | 4 - 4 | 1497 | 123 | |
M13445.1 | - | 4 - 4 | 1464 | 123 | |
M28729.1 | - | 1 - 1 | 657 | 123 |