Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Spin1
Entrez Gene ID
20729
Gene Name
spindlin 1
Gene Aliases
Spin
Chromosome Location
Chr.13: 51099834 - 51152562 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001283028.1 NP_001269957.1 6 - 6 1097 102
NM_001283029.1 NP_001269958.1 6 - 6 962 102
NM_001283030.1 NP_001269959.1 5 - 5 821 102
NM_011462.3 NP_035592.1 4 - 4 726 102
NM_146043.4 NP_666155.1 6 - 6 1022 102
GenBank mRNA AK088127.1 - 6 - 6 963 102
AK134259.1 - 6 - 6 985 102
AK135816.1 - 4 - 4 721 102
AK135847.1 - 4 - 4 725 102
AK135933.1 - 4 - 4 726 102
AK136251.1 - 4 - 4 725 102
AK139899.1 - 4 - 4 723 102
AK162171.1 - 4 - 4 723 102
AK162195.1 - 4 - 4 723 102
AK162258.1 - 4 - 4 720 102
AK163263.1 - 4 - 4 722 102
AK163293.1 - 4 - 4 723 102
AK163308.1 - 4 - 4 726 102
AK163382.1 - 4 - 4 723 102
AK166528.1 - 4 - 4 726 102
AK166580.1 - 4 - 4 685 102
AK166677.1 - 4 - 4 722 102
AK168544.1 - 6 - 6 1023 102
BC016517.1 - 6 - 6 1008 102
U48972.1 - 4 - 4 720 102

View Full Product Details