The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146292.1 | NP_001139764.1 | 2 - 3 | 728 | 103 |
NM_001146293.1 | NP_001139765.1 | 2 - 3 | 728 | 103 | |
NM_001146294.1 | NP_001139766.1 | 2 - 3 | 728 | 103 | |
NM_001174074.1 | NP_001167545.1 | 2 - 3 | 728 | 103 | |
NM_133195.3 | NP_573458.2 | 2 - 3 | 728 | 103 | |
GenBank mRNA | AF515450.1 | - | 2 - 3 | 496 | 103 |
AK048427.1 | - | 2 - 3 | 905 | 103 | |
AK051458.1 | - | 2 - 3 | 449 | 103 | |
BC048405.1 | - | 2 - 3 | 728 | 103 | |
BC052744.1 | - | 2 - 3 | 728 | 103 |