The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083315.2 | NP_001076784.1 | 4 - 5 | 744 | 72 |
NM_001289624.1 | NP_001276553.1 | 4 - 5 | 668 | 72 | |
NM_001289625.1 | NP_001276554.1 | 4 - 5 | 668 | 72 | |
NM_001289626.1 | NP_001276555.1 | 4 - 5 | 668 | 72 | |
NM_001289627.1 | NP_001276556.1 | 4 - 5 | 402 | 72 | |
NM_001289629.1 | NP_001276558.1 | 4 - 5 | 402 | 72 | |
NM_022332.3 | NP_071727.2 | 4 - 5 | 744 | 72 | |
GenBank mRNA | AF234884.1 | - | 4 - 5 | 695 | 72 |
AF234885.1 | - | 4 - 5 | 695 | 72 | |
AK079107.1 | - | 4 - 5 | 745 | 72 | |
AK132103.1 | - | 4 - 5 | 698 | 72 | |
AK151550.1 | - | 4 - 5 | 699 | 72 | |
AY007792.1 | - | 4 - 5 | 630 | 72 | |
AY007793.1 | - | 4 - 5 | 630 | 72 | |
AY007794.1 | - | 5 - 6 | 659 | 72 | |
AY007795.1 | - | 5 - 6 | 659 | 72 | |
AY007796.1 | - | 5 - 6 | 469 | 72 | |
AY007797.1 | - | 5 - 6 | 469 | 72 | |
AY007798.1 | - | 4 - 5 | 402 | 72 | |
AY007799.1 | - | 4 - 5 | 402 | 72 | |
AY551821.1 | - | 4 - 5 | 395 | 72 | |
AY551822.1 | - | 4 - 5 | 395 | 72 | |
BC024652.1 | - | 4 - 5 | 668 | 72 | |
BC051429.1 | - | 4 - 5 | 522 | 72 | |
BC060630.1 | - | 4 - 5 | 585 | 72 | |
BC108418.1 | - | 4 - 5 | 688 | 72 | |
CT010245.1 | - | 4 - 5 | 321 | 72 |