Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hepacam2
Entrez Gene ID
101202
Gene Name
HEPACAM family member 2
Gene Aliases
AI987662
Chromosome Location
Chr.6: 3457089 - 3498189 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178899.5 NP_849230.1 9 - 10 1406 149
GenBank mRNA AK033728.1 - 9 - 10 1408 149
AK050501.1 - 9 - 10 1407 149
BC096374.1 - 9 - 10 1393 149

View Full Product Details