The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172307.3 | NP_758511.1 | 10 - 11 | 1471 | 68 |
GenBank mRNA | AK077220.1 | - | - | 1417 | 68 |
AK148325.1 | - | 10 - 11 | 1436 | 68 | |
AK164486.1 | - | 10 - 11 | 1407 | 68 | |
BC038343.1 | - | 10 - 11 | 1398 | 68 |