Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lrrc16a
Entrez Gene ID
68732
Gene Name
leucine rich repeat containing 16A
Gene Aliases
1110037D04Rik, AI425970, CARMIL, D130057M20, Lrrc16
Chromosome Location
Chr.13: 24012481 - 24280790 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026825.3 NP_081101.3 19 - 20 1678 76
GenBank mRNA AK043591.1 - 4 - 5 258 76
AK049059.1 - 1 - 2 54 76
AY437876.1 - 19 - 20 1683 76
BC150819.1 - 18 - 19 1592 76

View Full Product Details