Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vwa7
Entrez Gene ID
27762
Gene Name
von Willebrand factor A domain containing 7
Gene Aliases
C6orf27, D17H6S56E-3, G7c, NG37
Chromosome Location
Chr.17: 35016357 - 35026741 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138582.1 NP_613048.1 14 - 15 2387 70
GenBank mRNA AF134319.1 - 14 - 15 2387 70
BC137792.1 - 14 - 15 2328 70
BC144911.1 - 14 - 15 2265 70

View Full Product Details