Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pmel
Entrez Gene ID
20431
Gene Name
premelanosome protein
Gene Aliases
D10H12S53E, D12S53Eh, Pmel17, Si, Silv, gp100, gp87
Chromosome Location
Chr.10: 128705168 - 128720238 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021882.4 NP_068682.2 2 - 3 218 80
GenBank mRNA AK012808.1 - 2 - 3 219 80
AK131956.1 - 2 - 3 220 80
BC075667.1 - 2 - 3 205 80
BC082555.1 - 2 - 3 204 80
U14133.1 - 2 - 3 193 80

View Full Product Details