Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc17a1
Entrez Gene ID
20504
Gene Name
solute carrier family 17 (sodium phosphate), member 1
Gene Aliases
NAPI-1, Napi1, Npt1
Chromosome Location
Chr.13: 23867723 - 23895730 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170638.1 NP_001164109.1 1 - 2 76 59
GenBank mRNA AF361761.1 - 1 - 2 59 59
AK164712.1 - 1 - 2 60 59
CJ065843.1 - 1 - 2 77 59

View Full Product Details