Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc17a2
Entrez Gene ID
218103
Gene Name
solute carrier family 17 (sodium phosphate), member 2
Gene Aliases
C730032N17Rik, NPT3
Chromosome Location
Chr.13: 23806941 - 23825191 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144836.2 NP_659085.1 5 - 6 968 65
GenBank mRNA AK040717.1 - 5 - 6 969 65
AK050184.1 - 5 - 6 951 65
BC018306.1 - 4 - 5 606 65

View Full Product Details