Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Clcn1
Entrez Gene ID
12723
Gene Name
chloride channel 1
Gene Aliases
Clc-1, Clc1, SMCC1, adr, mto, myotonia, nmf355
Chromosome Location
Chr.6: 42282593 - 42315759 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013491.2 NP_038519.1 1 - 2 256 78
GenBank mRNA AK036735.1 - 1 - 2 258 78
BC114336.1 - 1 - 2 221 78

View Full Product Details