Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Smarcd3
Entrez Gene ID
66993
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 3
Gene Aliases
1500001J14Rik, 2210409C08Rik, BAF60C
Chromosome Location
Chr.5: 24592622 - 24624456 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025891.3 NP_080167.3 7 - 8 927 69
GenBank mRNA AK005094.1 - 7 - 8 874 69
AK158019.1 - - 897 69
BC013122.1 - 7 - 8 964 69
BC060525.1 - 7 - 8 826 69

View Full Product Details