The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252292.1 | NP_001239221.1 | 6 - 7 | 841 | 63 |
NM_001252293.1 | NP_001239222.1 | 5 - 6 | 562 | 63 | |
NM_008590.2 | NP_032616.1 | 5 - 6 | 740 | 63 | |
GenBank mRNA | AF482999.1 | - | 6 - 7 | 562 | 63 |
AK031718.1 | - | 5 - 6 | 739 | 63 | |
AK032881.1 | - | 5 - 6 | 739 | 63 | |
AK034949.1 | - | 5 - 6 | 738 | 63 | |
AK146457.1 | - | 6 - 7 | 841 | 63 | |
AK168714.1 | - | 5 - 6 | 738 | 63 | |
AK168732.1 | - | 5 - 6 | 738 | 63 | |
AK168743.1 | - | 5 - 6 | 738 | 63 | |
AK168995.1 | - | 5 - 6 | 738 | 63 | |
AK169095.1 | - | 5 - 6 | 738 | 63 | |
AK169266.1 | - | 5 - 6 | 738 | 63 | |
BC006639.1 | - | 5 - 6 | 723 | 63 | |
CF916557.1 | - | 5 - 6 | 554 | 63 | |
D16262.1 | - | 5 - 6 | 727 | 63 |