The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136065.2 | NP_001129537.1 | 13 - 14 | 3179 | 72 |
NM_001294143.1 | NP_001281072.1 | 13 - 14 | 3258 | 72 | |
NM_001294144.1 | NP_001281073.1 | 13 - 14 | 3255 | 72 | |
NM_010433.2 | NP_034563.2 | 13 - 14 | 3339 | 72 | |
GenBank mRNA | AF077659.1 | - | 13 - 14 | 3102 | 72 |
AF170301.1 | - | 13 - 14 | 3008 | 72 | |
AF170302.1 | - | 13 - 14 | 3089 | 72 | |
AF208292.1 | - | 13 - 14 | 3092 | 72 | |
AF273680.1 | - | 13 - 14 | 2979 | 72 | |
AF333791.1 | - | 13 - 14 | 2882 | 72 | |
AF333792.1 | - | 13 - 14 | 2966 | 72 | |
BC031904.1 | - | 13 - 14 | 2991 | 72 |