Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Btn1a1
Entrez Gene ID
12231
Gene Name
butyrophilin, subfamily 1, member A1
Gene Aliases
Btn
Chromosome Location
Chr.13: 23456984 - 23465914 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013483.3 NP_038511.1 5 - 6 1025 87
GenBank mRNA AK142596.1 - 5 - 6 932 87
BC011497.1 - 5 - 6 999 87
BC031459.1 - 4 - 5 923 87
S80642.1 - 3 - 4 751 87

View Full Product Details