The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025537.3 | NP_079813.1 | 3 - 4 | 372 | 97 |
GenBank mRNA | AJ277891.1 | - | 2 - 3 | 209 | 97 |
AK009898.1 | - | 3 - 4 | 392 | 97 | |
AK012240.1 | - | 3 - 4 | 363 | 97 | |
AK018591.1 | - | 3 - 4 | 361 | 97 | |
AK020437.1 | - | 3 - 4 | 357 | 97 | |
AK169164.1 | - | 3 - 4 | 363 | 97 | |
AK171656.1 | - | 3 - 4 | 369 | 97 | |
AK172112.1 | - | 3 - 4 | 369 | 97 | |
AK215409.1 | - | 1 - 2 | 77 | 97 | |
BC034286.1 | - | 3 - 4 | 351 | 97 | |
BC057904.1 | - | 3 - 4 | 372 | 97 |