Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gdpd5
Entrez Gene ID
233552
Gene Name
glycerophosphodiester phosphodiesterase domain containing 5
Gene Aliases
BC024955, Gde2
Chromosome Location
Chr.7: 99381415 - 99460984 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201352.2 NP_958740.2 9 - 10 1455 70
GenBank mRNA AK141189.1 - 9 - 10 1488 70
AK154771.1 - 9 - 10 1592 70
BC024955.1 - 8 - 9 741 70
BC026428.1 - 5 - 6 388 70
BC082585.1 - 9 - 10 1455 70

View Full Product Details