Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ppme1
Entrez Gene ID
72590
Gene Name
protein phosphatase methylesterase 1
Gene Aliases
1110069N17Rik, 2700017M01Rik, PME-1, Pme1
Chromosome Location
Chr.7: 100326737 - 100371896 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028292.2 NP_082568.1 7 - 8 710 75
GenBank mRNA AK012256.1 - 7 - 8 782 75
AK028261.1 - 7 - 8 745 75
AK076992.1 - 7 - 8 1121 75
AK146051.1 - 7 - 8 692 75
AK146268.1 - 7 - 8 722 75
AK154876.1 - 7 - 8 783 75
BC014867.1 - 7 - 8 710 75
BC029064.1 - 7 - 8 698 75

View Full Product Details