The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198831.2 | NP_942128.2 | 5 - 6 | 493 | 63 |
NR_003559.1 | 5 - 6 | 458 | 63 | ||
GenBank mRNA | AK171917.1 | - | 5 - 6 | 466 | 63 |
BC037190.1 | - | 5 - 6 | 448 | 63 | |
BC096562.1 | - | 3 - 4 | 290 | 63 | |
BC116358.1 | - | 4 - 5 | 363 | 63 | |
BC116359.1 | - | 4 - 5 | 360 | 63 | |
BC128487.1 | - | 4 - 5 | 363 | 63 | |
BC128488.1 | - | 4 - 5 | 363 | 63 | |
BG077486.2 | - | 4 - 5 | 327 | 63 | |
BU604260.1 | - | 5 - 6 | 486 | 63 | |
CD363582.1 | - | 5 - 6 | 424 | 63 | |
DN174444.1 | - | 5 - 6 | 493 | 63 |