The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285870.1 | NP_001272799.1 | 2 - 3 | 363 | 62 |
NM_001285871.1 | NP_001272800.1 | 2 - 3 | 548 | 62 | |
NM_001285873.1 | NP_001272802.1 | 2 - 3 | 363 | 62 | |
NM_018880.3 | NP_061368.1 | 2 - 3 | 337 | 62 | |
GenBank mRNA | AB030912.1 | - | 2 - 3 | 364 | 62 |
AF220019.1 | - | 2 - 3 | 256 | 62 | |
AK019165.1 | - | 2 - 3 | 298 | 62 | |
AK144674.1 | - | 2 - 3 | 266 | 62 | |
AK170029.1 | - | 2 - 3 | 301 | 62 | |
BC003347.1 | - | 2 - 3 | 378 | 62 | |
BC034263.1 | - | 2 - 3 | 302 | 62 | |
CD348171.1 | - | 2 - 3 | 363 | 62 |