Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Umod
Entrez Gene ID
22242
Gene Name
uromodulin
Gene Aliases
THP, Urehd1, urehr4
Chromosome Location
Chr.7: 119462708 - 119479262 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009470.5 NP_033496.1 1 - 2 109 89
GenBank mRNA AK085460.1 - 1 - 2 103 89

View Full Product Details