The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163664.1 | NP_001157136.1 | 13 - 14 | 486 | 59 |
NM_001163665.1 | NP_001157137.1 | 13 - 14 | 486 | 59 | |
NM_001163666.1 | NP_001157138.1 | 11 - 12 | 459 | 59 | |
NM_001163667.1 | NP_001157139.1 | 12 - 13 | 474 | 59 | |
NM_001163668.1 | NP_001157140.1 | 11 - 12 | 435 | 59 | |
NM_001163669.1 | NP_001157141.1 | 12 - 13 | 447 | 59 | |
NM_001163670.1 | NP_001157142.1 | 9 - 10 | 402 | 59 | |
NM_011620.3 | NP_035750.2 | 10 - 11 | 414 | 59 | |
GenBank mRNA | AI173874.1 | - | 11 - 12 | 451 | 59 |
AI574395.1 | - | 13 - 14 | 480 | 59 | |
AI746973.1 | - | 11 - 12 | 454 | 59 | |
AK003147.1 | - | 9 - 10 | 369 | 59 | |
BC003747.1 | - | 13 - 14 | 482 | 59 | |
CB196352.1 | - | 3 - 4 | 238 | 59 | |
CK791812.1 | - | 2 - 3 | 124 | 59 | |
DV049834.1 | - | 10 - 11 | 461 | 59 | |
L48988.1 | - | 7 - 8 | 305 | 59 | |
L48989.1 | - | 7 - 8 | 305 | 59 | |
L48990.1 | - | 10 - 11 | 350 | 59 | |
L48991.1 | - | 7 - 8 | 332 | 59 | |
L48992.1 | - | 9 - 10 | 338 | 59 | |
L48993.1 | - | 9 - 10 | 338 | 59 | |
L49018.1 | - | 7 - 8 | 332 | 59 | |
U77779.1 | - | 10 - 11 | 407 | 59 |