Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gga2
Entrez Gene ID
74105
Gene Name
golgi associated, gamma adaptin ear containing, ARF binding protein 2
Gene Aliases
1200007E24Rik, mKIAA1080
Chromosome Location
Chr.7: 121986722 - 122021198 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028758.2 NP_083034.1 10 - 11 1125 79
GenBank mRNA AK004632.1 - 10 - 11 1097 79
AK034612.1 - 7 - 8 768 79
AK129280.1 - 10 - 11 1084 79
AK143691.1 - 10 - 11 1113 79
AK149798.1 - 10 - 11 1115 79
AK154904.1 - 10 - 11 1100 79
AK159953.1 - 10 - 11 1115 79
BC057377.1 - 3 - 4 628 79
BC062933.1 - 10 - 11 1094 79

View Full Product Details