Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gga2
Entrez Gene ID
74105
Gene Name
golgi associated, gamma adaptin ear containing, ARF binding protein 2
Gene Aliases
1200007E24Rik, mKIAA1080
Chromosome Location
Chr.7: 121986722 - 122021198 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028758.2 NP_083034.1 12 - 13 1233 69
GenBank mRNA AK004632.1 - 12 - 13 1205 69
AK034612.1 - 9 - 10 876 69
AK129280.1 - 12 - 13 1192 69
AK143691.1 - 12 - 13 1221 69
AK149798.1 - 12 - 13 1223 69
AK154904.1 - 12 - 13 1208 69
AK159953.1 - 12 - 13 1223 69
AK200704.1 - 2 - 3 122 69
AK218023.1 - 3 - 4 140 69
BC057377.1 - 5 - 6 736 69
BC062933.1 - 12 - 13 1202 69

View Full Product Details