The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171147.1 | NP_001164618.1 | 3 - 4 | 847 | 58 |
NM_009534.3 | NP_033560.1 | 3 - 4 | 847 | 58 | |
GenBank mRNA | AK182200.1 | - | 1 - 2 | 31 | 58 |
BC014733.1 | - | 3 - 4 | 844 | 58 | |
BC039125.1 | - | 3 - 4 | 702 | 58 | |
BC094313.1 | - | 3 - 4 | 835 | 58 | |
X80508.1 | - | 3 - 4 | 839 | 58 |