The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163276.1 | NP_001156748.1 | 5 - 6 | 992 | 81 |
NM_198107.2 | NP_932775.2 | 5 - 6 | 995 | 81 | |
GenBank mRNA | AK042340.1 | - | 5 - 6 | 957 | 81 |
AK138125.1 | - | 5 - 6 | 979 | 81 | |
AK149345.1 | - | 5 - 6 | 976 | 81 | |
AK191258.1 | - | 1 - 2 | 44 | 81 | |
AK213091.1 | - | 1 - 2 | 276 | 81 | |
BC057056.1 | - | 5 - 6 | 949 | 81 |