The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170935.1 | NP_001164406.1 | 4 - 5 | 1131 | 94 |
NM_181681.2 | NP_859009.2 | 4 - 5 | 1131 | 94 | |
NR_033210.1 | 3 - 4 | 1272 | 94 | ||
GenBank mRNA | AF541278.1 | - | 4 - 5 | 1131 | 94 |
AK083932.1 | - | 3 - 4 | 929 | 94 | |
AK144442.1 | - | 4 - 5 | 809 | 94 | |
AK220474.1 | - | 3 - 4 | 1208 | 94 | |
BC005764.1 | - | 4 - 5 | 886 | 94 | |
BC066006.1 | - | 4 - 5 | 726 | 94 | |
BC098099.1 | - | 4 - 5 | 884 | 94 |