Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Boc
Entrez Gene ID
117606
Gene Name
biregional cell adhesion molecule-related/down-regulated by oncogenes (Cdon) binding protein
Gene Aliases
4732455C11, mFLJ00376
Chromosome Location
Chr.16: 44485045 - 44558870 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172506.2 NP_766094.1 5 - 6 1183 96
GenBank mRNA AF388037.1 - 4 - 5 639 96
AK028770.1 - 5 - 6 1204 96
AK028889.1 - 5 - 6 1184 96
AK043379.1 - 5 - 6 1193 96
AK131182.1 - 1 - 2 126 96
BC062892.1 - 5 - 6 1163 96
BC078631.1 - 5 - 6 1130 96

View Full Product Details