The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007675.4 | NP_031701.3 | 3 - 4 | 853 | 112 |
GenBank mRNA | AK018529.1 | - | 3 - 4 | 833 | 112 |
BC003346.1 | - | 3 - 4 | 874 | 112 | |
BE949028.1 | - | 1 - 2 | 112 | ||
L38422.1 | - | 3 - 4 | 817 | 112 |