The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161853.1 | NP_001155325.1 | 8 - 9 | 1310 | 80 |
NM_011418.2 | NP_035548.1 | 8 - 9 | 1337 | 80 | |
GenBank mRNA | AB041578.1 | - | 8 - 9 | 1293 | 80 |
AJ011739.1 | - | 8 - 9 | 1311 | 80 | |
AJ011740.1 | - | 8 - 9 | 1311 | 80 | |
AK008558.1 | - | 6 - 7 | 1042 | 80 | |
AK075906.1 | - | 8 - 9 | 1320 | 80 | |
AK145552.1 | - | 8 - 9 | 1299 | 80 | |
AK147638.1 | - | 4 - 5 | 7126 | 80 | |
AK147685.1 | - | 4 - 5 | 7126 | 80 | |
AK150169.1 | - | 8 - 9 | 1311 | 80 | |
BC025163.1 | - | 8 - 9 | 1291 | 80 | |
BC035269.1 | - | 6 - 7 | 3468 | 80 |