Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Upb1
Entrez Gene ID
103149
Gene Name
ureidopropionase, beta
Gene Aliases
AI195023
Chromosome Location
Chr.10: 75406911 - 75441679 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133995.4 NP_598756.1 2 - 3 426 76
GenBank mRNA AK149373.1 - 2 - 3 323 76
AK149480.1 - 2 - 3 323 76
AK165429.1 - 2 - 3 357 76
BC021388.1 - 2 - 3 311 76

View Full Product Details