The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027422.2 | NP_081698.2 | 2 - 3 | 108 | 76 |
GenBank mRNA | AK154672.1 | - | 2 - 3 | 119 | 76 |
BC067013.1 | - | 2 - 3 | 108 | 76 |