Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rab8b
Entrez Gene ID
235442
Gene Name
RAB8B, member RAS oncogene family
Gene Aliases
5930437D16, D330025I23Rik
Chromosome Location
Chr.9: 66843664 - 66919705 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173413.3 NP_775589.1 5 - 6 487 140
GenBank mRNA AB232601.1 - 5 - 6 415 140
AK031251.1 - 5 - 6 434 140
AK084650.1 - 5 - 6 437 140
AK155621.1 - 5 - 6 437 140
BC059208.1 - 5 - 6 469 140
BC157974.1 - 5 - 6 435 140

View Full Product Details