The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018889.3 | NP_061377.2 | 11 - 12 | 1591 | 77 |
GenBank mRNA | AK153819.1 | - | 11 - 12 | 1508 | 77 |
BC052658.1 | - | 11 - 12 | 1551 | 77 | |
D84436.1 | - | 11 - 12 | 1591 | 77 |