Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fxyd2
Entrez Gene ID
11936
Gene Name
FXYD domain-containing ion transport regulator 2
Gene Aliases
Atp1g1
Chromosome Location
Chr.9: 45399709 - 45410278 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007503.3 NP_031529.2 5 - 6 274 144
NM_052823.2 NP_439888.1 5 - 6 347 144
NR_028499.1 5 - 6 479 144
GenBank mRNA AA396010.1 - 5 - 6 143
AI316264.1 - 5 - 6 258 144
AK002215.1 - 5 - 6 262 144
AK002228.1 - 5 - 6 275 144
AK007105.1 - 3 - 4 149 144
AY626243.1 - 5 - 6 396 144
AY626244.1 - 5 - 6 347 144
AY626245.1 - 5 - 6 432 144
BC024614.1 - 5 - 6 263 144
BC069892.1 - 4 - 5 1405 144
BU756853.1 - 1 - 2 144
CB600439.1 - 5 - 6 307 144

View Full Product Details