Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ulk3
Entrez Gene ID
71742
Gene Name
unc-51-like kinase 3
Gene Aliases
1200015E14Rik
Chromosome Location
Chr.9: 57589137 - 57596233 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027895.1 NP_082171.1 9 - 10 1134 56
GenBank mRNA AK004783.1 - 7 - 8 1716 56
AK154642.1 - 9 - 10 1047 56
BC037093.1 - 9 - 10 1155 56
BC039958.1 - 5 - 6 496 56
BC151153.1 - 9 - 10 1060 56

View Full Product Details