Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tex9
Entrez Gene ID
21778
Gene Name
testis expressed gene 9
Gene Aliases
Gm19407, tsec-1
Chromosome Location
Chr.9: 72458055 - 72510944 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009359.4 NP_033385.2 11 - 12 1091 90
GenBank mRNA AK008505.1 - 11 - 12 1089 90
AK032490.1 - 10 - 11 1199 90
BC006667.1 - 2 - 3 1739 90
BC141033.1 - 11 - 12 1233 90
BC145184.1 - 12 - 13 1332 90
BC145185.1 - 11 - 12 1233 90
BC145186.1 - 10 - 11 1059 90

View Full Product Details