The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173777.3 | NP_776138.2 | 5 - 6 | 727 | 78 |
GenBank mRNA | AK037199.1 | - | 5 - 6 | 742 | 78 |
BC115981.1 | - | 5 - 6 | 727 | 78 | |
BC117536.1 | - | 5 - 6 | 732 | 78 |