Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tmed1
Entrez Gene ID
17083
Gene Name
transmembrane emp24 domain containing 1
Gene Aliases
Il1rl1l, Ly84l, St2l
Chromosome Location
Chr.9: 21507380 - 21510187 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010744.3 NP_034874.2 2 - 3 288 61
GenBank mRNA AK133513.1 - 2 - 3 288 61
BC138498.1 - 2 - 3 336 61
BC138499.1 - 2 - 3 335 61
U41805.1 - 2 - 3 264 61

View Full Product Details