Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tbc1d30
Entrez Gene ID
74694
Gene Name
TBC1 domain family, member 30
Gene Aliases
4930505D03Rik
Chromosome Location
Chr.10: 121263819 - 121311189 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK015700.1 - 1 - 2 68 67

View Full Product Details