Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Srgap1
Entrez Gene ID
117600
Gene Name
SLIT-ROBO Rho GTPase activating protein 1
Gene Aliases
4930572H05Rik, Arhgap13
Chromosome Location
Chr.10: 121780991 - 122047315 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081037.2 NP_001074506.1 5 - 6 797 85
NM_001242411.1 NP_001229340.1 5 - 6 797 85
GenBank mRNA BC120892.1 - 5 - 6 790 85
BC120893.1 - 5 - 6 790 85
CB521559.1 - 4 - 5 506 85
CD351674.1 - 1 - 2 144 85

View Full Product Details